Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318521.1 | NP_001305450.1 | 20 | 2162 | ||
NM_006733.3 | NP_006724.2 | 19 | 2256 | |||
XM_005262111.2 | XP_005262168.1 | 20 | 2126 | |||
XM_011530894.2 | XP_011529196.1 | 20 | 2155 | |||
XM_011530895.2 | XP_011529197.1 | 21 | 2442 | |||
XM_011530897.2 | XP_011529199.1 | 21 | 2317 | |||
XM_011530899.2 | XP_011529201.1 | 19 | 2123 | |||
XM_017029383.1 | XP_016884872.1 | 20 | 2155 | |||
GenBank mRNA | AK302986.1 | - | 20 | 2155 | ||
BG114761.1 | - | 407 | ||||
X97249.1 | - | 19 | 2255 |