Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009322.3 | NP_033348.2 | 3 | 1208 | ||
GenBank mRNA | BC052737.1 | - | 3 | 1208 | 6825394 | |
BC058399.1 | - | 3 | 1112 | 6826831 | ||
U49251.1 | - | 3 | 1048 |