Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009324.2 | NP_033350.2 | 7 | 2307 | ||
GenBank mRNA | AK137225.1 | - | 7 | 1518 | ||
AK149348.1 | - | 7 | 2115 | |||
U15566.1 | - | 7 | 1843 |