Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006739.3 | NP_006730.2 | 12 | 1605 | ||
XM_006724242.3 | XP_006724305.1 | 12 | 1660 | |||
GenBank mRNA | AB209612.1 | - | 6 | 1093 | ||
AK122853.1 | - | 11 | 1453 | |||
AK130620.1 | - | 11 | 1559 | |||
AK223323.1 | - | 12 | 1595 | |||
AK291136.1 | - | 12 | 1586 | |||
AK298624.1 | - | 12 | 1482 | |||
AK301121.1 | - | 523 | ||||
AK308343.1 | - | 10 | 1206 | |||
AK312533.1 | - | 12 | 1585 | |||
BC000142.1 | - | 12 | 1557 | 2900229 | ||
BC003656.2 | - | 11 | 1531 | 3542201 | ||
CR456517.1 | - | 12 | 1545 | |||
D83986.1 | - | 12 | 1593 | |||
X74795.2 | - | 11 | 1533 |