Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB011550.1 | - | 10, 11 | 1166 | ||
AK076365.1 | - | 8, 9 | 2082 | |||
AK133474.1 | - | 10, 11 | 1134 | |||
AK139088.1 | - | 10, 11 | 1372 | |||
AK144793.1 | - | 10, 11 | 1350 | |||
AK155268.1 | - | 7, 8 | 2211 | |||
AK193704.1 | - | 3, 4 | 236 | |||
BC015077.1 | - | 10, 11 | 1048 | |||
BC145595.1 | - | 9, 10 | 1140 | |||
BC145596.1 | - | 10, 11 | 1055 | |||
U81490.1 | - | 10, 11 | 1106 |