Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006743.4 | NP_006734.1 | 6 | 687 | ||
GenBank mRNA | AK000859.1 | - | 6 | 556 | ||
AK026664.1 | - | 3 | 659 | |||
AK309137.1 | - | 6 | 945 | |||
AY203954.1 | - | 5 | 1149 | |||
BC006825.1 | - | 6 | 516 | 3449185 | ||
BQ429805.1 | - | 5 | 451 | |||
BX647914.1 | - | 1 | 2552 |