Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166585.1 | NP_001160057.1 | 2, 3 | 326 | ||
NM_009346.3 | NP_033372.1 | 3, 4 | 733 | |||
GenBank mRNA | AK148375.1 | - | 3, 4 | 699 | ||
BC052021.1 | - | 2, 3 | 267 | |||
BC060138.1 | - | 2, 3 | 267 | 6410573 | ||
L06865.1 | - | 2, 3 | 272 | |||
L13853.1 | - | 3, 4 | 460 | |||
S74227.1 | - | 3, 4 | 733 |