Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Terf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286628.1 | NP_001273557.1 | 6 | 833 | ||
NM_009352.3 | NP_033378.1 | 7 | 920 | |||
GenBank mRNA | AB109563.1 | - | 7 | 900 | ||
AK132022.1 | - | 6 | 816 | |||
BC034866.1 | - | 7 | 908 | 3470277 | ||
U65586.1 | - | 7 | 900 |