Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Terf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286200.1 | NP_001273129.1 | 11 | 1422 | ||
NM_009353.2 | NP_033379.2 | 10 | 1614 | |||
GenBank mRNA | AF003000.1 | - | 10 | 1508 | ||
AK089419.1 | - | 11 | 1397 | |||
AK160421.1 | - | 11 | 1090 |