Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009356.2 | NP_033382.2 | 3, 4 | 656 | ||
GenBank mRNA | AB008911.1 | - | 3, 4 | 639 | ||
AK006524.1 | - | 1, 2 | 266 | |||
AK132909.1 | - | 3, 4 | 686 | |||
BC145989.1 | - | 3, 4 | 671 | 40131292 |