Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009357.2 | NP_033383.1 | 3 | 380 | ||
GenBank mRNA | AK149789.1 | - | 3 | 345 | ||
AK157834.1 | - | 3 | 359 | |||
AK190569.1 | - | 1 | 14 | |||
BC003802.1 | - | 3 | 324 | 3486968 | ||
X81058.1 | - | 3 | 326 |