Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009359.4 | NP_033385.2 | 4 | 223 | ||
GenBank mRNA | AB000619.1 | - | 5 | 471 | ||
AK008505.1 | - | 4 | 221 | |||
AK012189.1 | - | 4 | 222 | |||
AK018568.1 | - | 221 | ||||
AK032490.1 | - | 3 | 330 | |||
AK141174.1 | - | 3 | 333 | |||
BC141033.1 | - | 4 | 365 | 9055645 | ||
BC145184.1 | - | 4 | 365 | |||
BC145185.1 | - | 4 | 365 | 9053736 | ||
BC145186.1 | - | 4 | 365 | 9053737 |