Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009360.4 | NP_033386.1 | 4 | 466 | ||
GenBank mRNA | AK004857.1 | - | 4 | 464 | ||
AK017974.1 | - | 4 | 493 | |||
AK050446.1 | - | 4 | 467 | |||
AK161747.1 | - | 4 | 493 | |||
AK163563.1 | - | 4 | 484 | |||
AK167348.1 | - | 4 | 467 | |||
AK167777.1 | - | 4 | 493 | |||
AK169808.1 | - | 4 | 494 | |||
AK187847.1 | - | 1 | 66 | |||
BC001987.1 | - | 4 | 461 | 3487433 | ||
BC083084.1 | - | 4 | 461 | 6514962 | ||
L07107.1 | - | 3 | 263 | |||
U57939.1 | - | 4 | 458 | |||
U63858.1 | - | 4 | 300 | |||
U63859.1 | - | 4 | 666 | |||
U63860.1 | - | 2 | 94 |