Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009360.4 | NP_033386.1 | 5, 6 | 646 | ||
GenBank mRNA | AK004857.1 | - | 5, 6 | 644 | ||
AK017974.1 | - | 5, 6 | 674 | |||
AK050446.1 | - | 5, 6 | 647 | |||
AK161747.1 | - | 5, 6 | 673 | |||
AK163563.1 | - | 5, 6 | 664 | |||
AK167348.1 | - | 5, 6 | 647 | |||
AK167777.1 | - | 5, 6 | 673 | |||
AK169808.1 | - | 5, 6 | 674 | |||
AK178048.1 | - | 2, 3 | 146 | |||
AK187847.1 | - | 2, 3 | 247 | |||
AK213712.1 | - | 2, 3 | 221 | |||
BC001987.1 | - | 5, 6 | 641 | 3487433 | ||
BC083084.1 | - | 5, 6 | 641 | 6514962 | ||
L07107.1 | - | 4, 5 | 443 | |||
U57939.1 | - | 5, 6 | 638 | |||
U63858.1 | - | 5, 6 | 480 | |||
U63859.1 | - | 5, 6 | 846 | |||
U63860.1 | - | 3, 4 | 274 |