Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSFA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130445.2 | NP_001123917.1 | 4 | 687 | ||
NM_001287503.1 | NP_001274432.1 | 4 | 687 | |||
NM_001287504.1 | NP_001274433.1 | 3 | 637 | |||
NM_006751.6 | NP_006742.2 | 4 | 687 | |||
NR_109843.1 | 4 | 687 | ||||
XM_005246812.1 | XP_005246869.1 | 4 | 595 | |||
XM_005246813.1 | XP_005246870.1 | 4 | 595 | |||
XM_011511701.2 | XP_011510003.1 | 4 | 595 | |||
XM_017004782.1 | XP_016860271.1 | 4 | 595 | |||
GenBank mRNA | AB067514.2 | - | 4 | 535 | ||
AB116937.1 | - | 4 | 402 | |||
AK291745.1 | - | 4 | 423 | |||
AK304238.1 | - | 3 | 157 | |||
BC028706.2 | - | 4 | 448 | 4823606 | ||
BC052581.1 | - | 4 | 456 | 6084501 | ||
BC064499.1 | - | 4 | 456 | 6251533 | ||
BX647257.1 | - | 4 | 418 | |||
BX647967.1 | - | 4 | 320 | |||
BX648888.1 | - | 4 | 208 | |||
CR749488.1 | - | 3 | 432 |