Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278942.1 | NP_001265871.1 | 5 | 1974 | ||
NM_006753.5 | NP_006744.2 | 5 | 1975 | |||
NR_103874.1 | 5 | 1922 | ||||
GenBank mRNA | AK054884.1 | - | 5 | 1972 | ||
BC003001.1 | - | 5 | 1902 | |||
BC006197.2 | - | 5 | 1902 | 2820773 | ||
CB158198.1 | - | 1 | 98 |