Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173505.1 | NP_001166976.1 | 4, 5 | 466 | ||
NM_001289894.1 | NP_001276823.1 | 4, 5 | 466 | |||
NM_001289896.1 | NP_001276825.1 | 4, 5 | 466 | |||
NM_009379.3 | NP_033405.1 | 4, 5 | 620 | |||
GenBank mRNA | AI314551.1 | - | 4, 5 | 466 | ||
AK047512.1 | - | 4, 5 | 621 | |||
BC003803.1 | - | 4, 5 | 732 | 3593885 | ||
L34169.1 | - | 4, 5 | 402 |