Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009380.3 | NP_033406.1 | 1 | 418 | ||
GenBank mRNA | AB221613.1 | - | 1 | 185 | ||
BC089035.1 | - | 1 | 418 | 30695885 | ||
BC119552.1 | - | 1 | 355 | 40057420 | ||
BC119553.1 | - | 1 | 359 | 40057421 | ||
S62758.1 | - | 1 | 292 |