Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tiam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145886.1 | NP_001139358.1 | 4 | 774 | ||
NM_009384.3 | NP_033410.2 | 4 | 809 | |||
GenBank mRNA | AK147392.1 | - | 4 | 775 | ||
BC065126.1 | - | 5 | 795 | 30534934 | ||
U05245.1 | - | 4 | 812 |