Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271729.1 | NP_001258658.1 | 6 | 883 | ||
NM_009387.2 | NP_033413.2 | 7 | 915 | |||
GenBank mRNA | AK085188.1 | - | 8 | 764 | ||
AK087988.1 | - | 7 | 718 | |||
AK139244.1 | - | 7 | 735 | |||
BC012215.1 | - | 7 | 719 | 3969287 | ||
BC091758.1 | - | 7 | 748 | 4978985 | ||
M11945.1 | - | 7 | 712 | |||
M19438.1 | - | 6 | 605 | |||
S57244.1 | - | 7 | 712 |