Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009388.6 | NP_033414.1 | 2 | 346 | ||
GenBank mRNA | AF195533.1 | - | 1 | 12 | ||
AK002627.1 | - | 2 | 299 | |||
AK012794.1 | - | 2 | 301 | |||
AK030446.1 | - | 2 | 329 | |||
AK050793.1 | - | 2 | 297 | |||
AK140965.1 | - | 2 | 330 | |||
AK144146.1 | - | 2 | 301 | |||
AK145677.1 | - | 2 | 299 | |||
AK146157.1 | - | 2 | 299 | |||
AK150139.1 | - | 2 | 299 | |||
AK150769.1 | - | 2 | 295 | |||
AK150844.1 | - | 2 | 295 | |||
AK150856.1 | - | 2 | 295 | |||
AK150901.1 | - | 2 | 299 | |||
AK151916.1 | - | 2 | 295 | |||
AK151969.1 | - | 2 | 295 | |||
AK152245.1 | - | 2 | 295 | |||
AK152460.1 | - | 2 | 315 | |||
AK152886.1 | - | 2 | 295 | |||
AK159922.1 | - | 2 | 299 | |||
AK166763.1 | - | 2 | 333 | |||
AK167084.1 | - | 2 | 333 | |||
AK181952.1 | - | 1 | 111 | |||
BC055336.1 | - | 2 | 273 | 6413263 | ||
BY276711.1 | - | 2 | 347 |