Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009418.2 | NP_033444.1 | 11 | 1372 | ||
GenBank mRNA | AK087835.1 | - | 11 | 1356 | ||
AK141908.1 | - | 11 | 1356 | |||
AK154144.1 | - | 11 | 1351 | |||
AK159865.1 | - | 11 | 1364 | |||
AK171519.1 | - | 11 | 1366 | |||
BC011275.1 | - | 11 | 1346 | |||
BC052318.1 | - | 11 | 1303 | |||
BC058239.1 | - | 11 | 1311 | 6308147 | ||
BC060627.1 | - | 11 | 1378 | |||
X81323.1 | - | 11 | 1373 |