Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crisp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204071.1 | NP_001191000.1 | 5 | 397 | ||
NM_009420.2 | NP_033446.1 | 5 | 415 | |||
GenBank mRNA | AK018856.1 | - | 5 | 346 | ||
BC049615.1 | - | 5 | 367 | 6742033 | ||
CJ046590.1 | - | 5 | 338 | |||
M25533.1 | - | 5 | 367 |