Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006770.3 | NP_006761.1 | 15, 16 | 1383 | ||
XM_011512082.1 | XP_011510384.1 | 15, 16 | 1386 | |||
XM_011512083.2 | XP_011510385.1 | 12, 13 | 995 | |||
GenBank mRNA | AF035819.1 | - | 15, 16 | 1251 | ||
AK297030.1 | - | 15, 16 | 1230 | |||
AK301407.1 | - | 16, 17 | 1400 | |||
AK313139.1 | - | 15, 16 | 1332 | |||
AX429246.1 | - | 15, 16 | 1251 | |||
BC016004.1 | - | 15, 16 | 1387 | 4618447 |