Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290413.1 | NP_001277342.1 | 5, 6 | 667 | ||
NM_009422.3 | NP_033448.2 | 5, 6 | 646 | |||
GenBank mRNA | AF027570.1 | - | 4, 5 | 558 | ||
AK038136.1 | - | 5, 6 | 595 | |||
AK052934.1 | - | 5, 6 | 595 | |||
AK151653.1 | - | 5, 6 | 584 | |||
AK152174.1 | - | 5, 6 | 614 | |||
AK159581.1 | - | 5, 6 | 616 | |||
AK159585.1 | - | 593 | ||||
BC003801.1 | - | 4, 5 | 572 | 3492839 | ||
L35303.1 | - | 5, 6 | 583 |