Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006773.3 | NP_006764.3 | 14 | 2189 | ||
GenBank mRNA | AB209392.1 | - | 5 | 4105 | ||
AK001467.1 | - | 14 | 2161 | |||
AK091227.1 | - | 12 | 1616 | |||
BC001238.1 | - | 14 | 2118 | 3452935 | ||
BC003360.1 | - | 14 | 2148 | 2900252 | ||
BC024739.1 | - | 14 | 2163 | 4995871 | ||
X98743.1 | - | 13 | 1927 |