Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009428.2 | NP_033454.1 | 6 | 2047 | ||
GenBank mRNA | AF029983.1 | - | 6 | 1702 | ||
AF060107.1 | - | 6 | 2046 | |||
AJ006204.1 | - | 5 | 1644 | |||
AK038531.1 | - | 6 | 1871 | |||
AK047867.1 | - | 4 | 1068 | |||
BC112972.1 | - | 6 | 1865 | 40049027 |