Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199219.1 | NP_001186148.1 | 3 | 2228 | ||
NM_006774.4 | NP_006765.4 | 3 | 2231 | |||
GenBank mRNA | AF128846.1 | - | 3 | 2236 | ||
AF128847.1 | - | 3 | 2236 | |||
BC033813.1 | - | 3 | 2222 | |||
BC103712.1 | - | 4 | 2169 |