Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009439.1 | NP_033465.1 | 5 | 957 | ||
GenBank mRNA | AK031366.1 | - | 5 | 947 | ||
AK077252.1 | - | 5 | 906 | |||
AK089933.1 | - | 5 | 915 | |||
AK145467.1 | - | 5 | 947 | |||
AK145488.1 | - | 5 | 946 | |||
AK153505.1 | - | 5 | 874 | |||
AK164333.1 | - | 5 | 921 | |||
AK164593.1 | - | 5 | 946 | |||
AK165520.1 | - | 5 | 892 | |||
AK166683.1 | - | 5 | 917 | |||
AK166714.1 | - | 5 | 945 | |||
AK167005.1 | - | 5 | 932 | |||
AK167285.1 | - | 5 | 912 | |||
AK169690.1 | - | 5 | 946 | |||
BC003197.1 | - | 5 | 957 | 3586427 |