Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009439.1 | NP_033465.1 | 5 | 1010 | ||
GenBank mRNA | AK031366.1 | - | 5 | 1000 | ||
AK077252.1 | - | 5 | 959 | |||
AK089933.1 | - | 5 | 968 | |||
AK145467.1 | - | 5 | 1000 | |||
AK145488.1 | - | 5 | 999 | |||
AK153505.1 | - | 5 | 927 | |||
AK164333.1 | - | 5 | 974 | |||
AK164593.1 | - | 5 | 999 | |||
AK165520.1 | - | 5 | 945 | |||
AK166683.1 | - | 5 | 970 | |||
AK166714.1 | - | 5 | 998 | |||
AK167005.1 | - | 5 | 985 | |||
AK167285.1 | - | 5 | 965 | |||
AK169690.1 | - | 5 | 999 | |||
BC003197.1 | - | 5 | 1010 | 3586427 |