Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136086.2 | NP_001129558.1 | 13 | 1680 | ||
NM_001291455.1 | NP_001278384.1 | 13 | 1930 | |||
NM_009468.5 | NP_033494.1 | 13 | 1861 | |||
GenBank mRNA | AK077000.1 | - | 13 | 1671 | ||
AK133040.1 | - | 13 | 1680 | |||
AK149024.1 | - | 13 | 1698 | |||
AK161509.1 | - | 13 | 1602 | |||
AK171653.1 | - | 6 | 761 | |||
BC023003.1 | - | 13 | 1545 | 5367381 | ||
X87817.1 | - | 13 | 1821 |