Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009481.2 | NP_033507.2 | 6 | 873 | ||
GenBank mRNA | AK028443.1 | - | 6 | 858 | ||
AK049283.1 | - | 7 | 986 | |||
DQ086491.1 | - | 6 | 834 | |||
U67874.1 | - | 6 | 834 | |||
Z78153.1 | - | 1 | 28 |