Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009482.2 | NP_033508.1 | 2 | 932 | ||
GenBank mRNA | AK010280.1 | - | 2 | 960 | ||
AK010589.1 | - | 2 | 966 | |||
AK131968.1 | - | 2 | 1002 | |||
AY606111.1 | - | 2 | 966 | |||
BC138841.1 | - | 2 | 941 | 8861863 | ||
BC138842.1 | - | 2 | 941 | 8861864 | ||
D31647.1 | - | 2 | 932 |