Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFG3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006796.2 | NP_006787.2 | 12 | 1646 | ||
XM_011525601.2 | XP_011523903.1 | 12 | 1679 | |||
GenBank mRNA | AK129504.1 | - | 5 | 472 | ||
BC024282.1 | - | 13 | 1499 | |||
BC065016.1 | - | 12 | 1568 | 6169804 | ||
Y18314.1 | - | 12 | 1565 |