Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vegfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025250.3 | NP_001020421.2 | 3, 4 | 1325 | ||
NM_001025257.3 | NP_001020428.2 | 3, 4 | 1325 | |||
NM_001287056.1 | NP_001273985.1 | 3, 4 | 1325 | |||
NM_001287057.1 | NP_001273986.1 | 3, 4 | 1325 | |||
NM_001287058.1 | NP_001273987.1 | 3, 4 | 1325 | |||
NM_009505.4 | NP_033531.3 | 3, 4 | 1325 | |||
GenBank mRNA | AY120866.1 | - | 1, 2 | 203 | ||
AY707864.1 | - | 4, 5 | 522 | |||
AY750956.1 | - | 4, 5 | 483 | |||
AY750957.1 | - | 4, 5 | 446 | |||
AY756068.1 | - | 3, 4 | 261 | |||
BC061468.1 | - | 3, 4 | 698 | 6816435 | ||
BQ832724.1 | - | 2, 3 | 226 | |||
CA321456.1 | - | 3 | 698 | |||
EF028705.1 | - | 4, 5 | 518 | |||
M95200.1 | - | 3, 4 | 390 | |||
S37052.1 | - | 3, 4 | 316 | |||
S38083.1 | - | 3, 4 | 316 | |||
S38100.1 | - | 3, 4 | 316 |