Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Was | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009515.2 | NP_033541.1 | 5, 6 | 567 | ||
GenBank mRNA | U29673.1 | - | 5, 6 | 528 | ||
U42471.1 | - | 5, 6 | 528 | |||
U54788.1 | - | 5, 6 | 528 |