Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009517.2 | NP_033543.2 | 2, 3 | 497 | ||
GenBank mRNA | AF012923.1 | - | 2, 3 | 451 | ||
BC138740.1 | - | 1, 2 | 316 | 8861762 | ||
BC138742.1 | - | 1, 2 | 316 | 8861764 | ||
BQ895699.1 | - | 2, 3 | 510 |