Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285792.1 | NP_001272721.1 | 7 | 1926 | ||
NM_001285794.1 | NP_001272723.1 | 6 | 1684 | |||
NM_001285795.1 | NP_001272724.1 | 5 | 1528 | |||
NM_009519.2 | NP_033545.1 | 5 | 1692 | |||
GenBank mRNA | AK031573.1 | - | 5 | 1536 | ||
AK077828.1 | - | 6 | 1625 | |||
AK138967.1 | - | 6 | 1685 | |||
X70800.1 | - | 5 | 1698 |