Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256224.1 | NP_001243153.1 | 4, 5 | 697 | ||
NM_009524.3 | NP_033550.2 | 4, 5 | 1320 | |||
GenBank mRNA | AK031512.1 | - | 4, 5 | 1030 | ||
AK031597.1 | - | 4, 5 | 1293 | |||
AK036824.1 | - | 4, 5 | 698 | |||
BC018425.1 | - | 4, 5 | 937 | 3487288 | ||
M89798.1 | - | 4, 5 | 1231 |