Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130914.1 | NP_001124386.1 | 5 | 811 | ||
NM_006806.4 | NP_006797.3 | 4 | 679 | |||
XM_011529441.1 | XP_011527743.1 | 6 | 919 | |||
XM_017028265.1 | XP_016883754.1 | 5 | 817 | |||
XM_017028266.1 | XP_016883755.1 | 5 | 774 | |||
GenBank mRNA | AK289408.1 | - | 4 | 627 | ||
BC011957.1 | - | 5 | 705 | 3859781 | ||
BT007276.1 | - | 4 | 551 | |||
CR457061.1 | - | 3 | 419 | |||
D64110.1 | - | 4 | 513 | |||
DR155504.1 | - | 4 | 595 |