Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291024.1 | NP_001277953.1 | 7 | 994 | ||
NM_009548.3 | NP_033574.2 | 8 | 1063 | |||
GenBank mRNA | AB013097.1 | - | 7 | 884 | ||
AK131651.1 | - | 7 | 938 | |||
BC013139.1 | - | 8 | 957 | 4500921 | ||
BC037118.1 | - | 8 | 1011 | 4506141 |