Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017956.2 | NP_001017956.1 | 13 | 1907 | ||
NM_001017957.2 | NP_001017957.1 | 13 | 1862 | |||
NM_001017958.2 | NP_001017958.1 | 14 | 2027 | |||
NM_001261420.1 | NP_001248349.1 | 13 | 1910 | |||
NM_001261421.1 | NP_001248350.1 | 12 | 1811 | |||
NM_001261422.1 | NP_001248351.1 | 12 | 1751 | |||
NM_001261423.1 | NP_001248352.1 | 12 | 1685 | |||
NM_006812.3 | NP_006803.1 | 14 | 2072 | |||
XM_005268581.1 | XP_005268638.1 | 14 | 2007 | |||
XM_006719200.1 | XP_006719263.1 | 14 | 1962 | |||
XM_006719201.1 | XP_006719264.1 | 13 | 1797 | |||
GenBank mRNA | AB002806.1 | - | 14 | 1906 | ||
AK130733.1 | - | 13 | 1720 | |||
AK291374.1 | - | 13 | 1789 | |||
AK293435.1 | - | 12 | 1497 | |||
AK296770.1 | - | 14 | 1837 | |||
AK298532.1 | - | 12 | 1621 | |||
AK303291.1 | - | 13 | 2259 | |||
AK303858.1 | - | 12 | 1561 | |||
AK304533.1 | - | 8 | 1099 | |||
AL137691.1 | - | 9 | 3083 | |||
BC000532.2 | - | 13 | 1712 | 2964645 | ||
BC006506.2 | - | 7 | 985 | |||
BC007254.1 | - | 13 | 1712 | 3027883 | ||
BC023513.2 | - | 13 | 1702 | 4123782 | ||
CR456791.1 | - | 13 | 1700 | |||
U41635.1 | - | 14 | 1950 |