Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290350.1 | NP_001277279.1 | 2, 3 | 311 | ||
NM_001290351.1 | NP_001277280.1 | 2, 3 | 302 | |||
NM_001290353.1 | NP_001277282.1 | 2, 3 | 209 | |||
NM_009554.4 | NP_033580.3 | 2, 3 | 299 | |||
GenBank mRNA | AK032469.1 | - | 2, 3 | 181 | ||
BC063757.1 | - | 2, 3 | 183 | 6331110 | ||
CF915126.1 | - | 2, 3 | 209 | |||
CJ078183.1 | - | 2, 3 | 299 | |||
CJ080611.1 | - | 2, 3 | 299 | |||
CO430987.1 | - | 2, 3 | 181 | |||
CO434318.1 | - | 2, 3 | 136 | |||
X64413.1 | - | 2, 3 | 105 | |||
X89264.1 | - | 3, 4 | 211 |