Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009569.3 | NP_033595.1 | 10 | 2122 | ||
GenBank mRNA | AF006492.1 | - | 10 | 2126 | ||
BC020344.1 | - | 1 | 365 |