Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009580.2 | NP_033606.2 | 2 | 308 | ||
GenBank mRNA | BC100347.1 | - | 2 | 326 | ||
BC125613.1 | - | 2 | 308 | 40130128 | ||
BC125615.1 | - | 2 | 308 | 40130130 | ||
U20448.1 | - | 2 | 339 |