Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163643.1 | NP_001157115.1 | 11 | 1815 | ||
NM_009582.4 | NP_033608.3 | 9 | 1639 | |||
GenBank mRNA | AK029882.1 | - | 9 | 1448 | ||
AK034374.1 | - | 11 | 1631 | |||
AK081623.1 | - | 4 | 307 | |||
AK166435.1 | - | 11 | 1735 | |||
BC047158.1 | - | 8 | 1433 | 6494216 | ||
BC057572.1 | - | 11 | 1639 | 6401321 | ||
U14636.1 | - | 9 | 1471 | |||
U23789.1 | - | 8 | 1429 |