Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tagln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011526.5 | NP_035656.1 | 4 | 668 | ||
GenBank mRNA | AK002880.1 | - | 4 | 507 | ||
AK077235.1 | - | 4 | 509 | |||
BC003795.1 | - | 4 | 497 | 3600413 | ||
L41154.1 | - | 4 | 507 | |||
U36588.1 | - | 4 | 507 |