Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011528.4 | NP_035658.1 | 6 | 865 | ||
GenBank mRNA | AK142030.1 | - | 6 | 823 | ||
BC003970.1 | - | 6 | 798 | |||
BC004754.1 | - | 6 | 820 | 3584511 | ||
BC094277.1 | - | 6 | 814 | 5008399 | ||
U63159.1 | - | 3 | 389 | |||
U67611.1 | - | 6 | 808 |