Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004367.5 | NP_004358.2 | 3 | 450 | ||
NM_031409.3 | NP_113597.2 | 3 | 713 | |||
GenBank mRNA | AK223494.1 | - | 3 | 369 | ||
AK293015.1 | - | 3 | 387 | |||
AY242126.1 | - | 1 | 145 | |||
BC037960.1 | - | 3 | 351 | 5748098 | ||
DN996219.1 | - | 3 | 438 | |||
U60000.1 | - | 1 | 145 | |||
U68030.1 | - | 4 | 409 |