Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006820.3 | NP_006811.2 | 9 | 1823 | ||
XM_005270391.3 | XP_005270448.1 | 8 | 1330 | |||
XM_005270392.4 | XP_005270449.1 | 7 | 1110 | |||
XM_005270393.4 | XP_005270450.1 | 6 | 1032 | |||
XM_006710304.3 | XP_006710367.1 | 8 | 1137 | |||
XM_011540539.2 | XP_011538841.1 | 9 | 1661 | |||
XM_017000120.1 | XP_016855609.1 | 9 | 1331 | |||
GenBank mRNA | AB000115.1 | - | 9 | 1747 | ||
AK223087.1 | - | 9 | 1757 | |||
AK293385.1 | - | 9 | 1422 | |||
AK303187.1 | - | 6 | 1024 | |||
AK316232.1 | - | 8 | 1272 | |||
AL832618.2 | - | 7 | 4426 | |||
BC015932.2 | - | 9 | 1729 | 3909331 |